Print…

Anti-GPR171 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB100926-100 ORB100926-200
BIRBORB100926-100EA 0 EUR
BIRBORB100926-100 BIRBORB100926-200
Anti-GPR171 Rabbit Polyclonal Antibody
Anticorpos
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: GPR171 (G protein-coupled receptor 171)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR