Print…

Anti-Destrin 19 Rabbit polyclonal antibody

Fornecedor: Biorbyt
ORB100163-100 ORB100163-200
BIRBORB100163-100EA 0 EUR
BIRBORB100163-100 BIRBORB100163-200
Anti-Destrin 19 Rabbit polyclonal antibody
Anticorpos
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: Destrin 19
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR