Print…

Anti-XRCC6 Goat Polyclonal Antibody

Fornecedor: Biorbyt
ORB19544-100 ORB19544-20
BIRBORB19544-100EA 0 EUR
BIRBORB19544-100 BIRBORB19544-20
Anti-XRCC6 Goat Polyclonal Antibody
Anticorpos
ELISA: 1:2000; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: XRCC6 (X-ray repair complementing defective repair in Chinese hamster cells 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR