Print…

Anti-NQO1 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB13617-100 ORB13617-200
BIRBORB13617-100EA 0 EUR
BIRBORB13617-100 BIRBORB13617-200
Anti-NQO1 Rabbit Polyclonal Antibody
Anticorpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: NQO1 (NAD(P)H dehydrogenase, quinone 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR