Print…
Anti-Dectin 1 Rabbit polyclonal antibody
Fornecedor: Biorbyt
ORB13296-100 ORB13296-200
BIRBORB13296-100EA 0 EUR
BIRBORB13296-100 BIRBORB13296-200
Anti-Dectin 1 Rabbit polyclonal antibody
Anticorpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: Dectin 1 (dectin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat