Print…
Anti-MET Rabbit Polyclonal Antibody
Fornecedor: Biorbyt
ORB10422-100 ORB10422-200
BIRBORB10422-100EA 0 EUR
BIRBORB10422-100 BIRBORB10422-200
Anti-MET Rabbit Polyclonal Antibody
Anticorpos
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: MET (met proto-oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar