Print…

Anti-BTBD10 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB2452-100 ORB2452-200
BIRBORB2452-100EA 0 EUR
BIRBORB2452-100 BIRBORB2452-200
Anti-BTBD10 Rabbit Polyclonal Antibody
Anticorpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: BTBD10 (BTB (POZ) domain containing 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR