Print…

Anti-COL2 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB1609-100 ORB1609-200
BIRBORB1609-100EA 0 EUR
BIRBORB1609-100 BIRBORB1609-200
Anti-COL2 Rabbit Polyclonal Antibody
Anticorpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR