Print…

Anti-Proteasome 19S S3 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB1455-100 ORB1455-200
BIRBORB1455-100EA 0 EUR
BIRBORB1455-100 BIRBORB1455-200
Anti-Proteasome 19S S3 Rabbit Polyclonal Antibody
Anticorpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: Proteasome 19S S3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR