Print…

Anti-SDHC Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB1149-100 ORB1149-200
BIRBORB1149-100EA 0 EUR
BIRBORB1149-100 BIRBORB1149-200
Anti-SDHC Rabbit Polyclonal Antibody
Anticorpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: SDHC (succinate dehydrogenase complex, subunit C, integral membrane protein, 15kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR