Print…
Capillary columns, Rxi®-5HT (fused silica)
Fornecedor: Restek
13906 13923 13909 13905 13924 13908
REST13906EA 591 EUR
REST13906 REST13923 REST13909 REST13905 REST13924 REST13908
Capillary columns, Rxi®-5HT (fused silica)
Cromatografia, Colunas
Fused capillary columns for high-temperature applications such as mineral oil extraction and analysis.
Low polarity phase; diphenyl dimethyl polysiloxane
Temperature range: –60 to +400 °C
Similar phases: DB-5ht, VF-5ht, ZB-5HTinferno.