Print…
Packaging kits, P650/P(I)620
Fornecedor: DAKLAPACK
520100 520125
DAKL520100EA 44.5 EUR
DAKL520100 DAKL520125
Packaging kits, P650/P(I)620
Embalagens para expedição
These kits contain a cardboard outer box (UN3373 or UN2814), a 500 ml green leak proof container, plastic shock absorbers and absorbing tube holders.
  • Non transparent
  • Made from cardboard
Just drop in your samples, seal the lid closed and fit back into the cardboard box and send with your preferred courier.