Print…
Safety shoes, lace-up, Protektor Line, 1602
Fornecedor: Abeba
1602/38 1602/46 1602/37 1602/39 1602/40 1602/41 1602/42 1602/43 1602/36 1602/47 1602/45 1602/48
ABEB1602/38EA 78.8 EUR
ABEB1602/38 ABEB1602/46 ABEB1602/37 ABEB1602/39 ABEB1602/40 ABEB1602/41 ABEB1602/42 ABEB1602/43 ABEB1602/36 ABEB1602/47 ABEB1602/45 ABEB1602/48
Safety shoes, lace-up, Protektor Line, 1602
Sapatos e botas
Anti-static shoes made from smooth leather upper, reflective material, and non slip PU sole.
  • Leather lining
  • Protective steel toecap (200 J)
  • PU sole with high slip resistance
Suitable for use in kitchens and in food processing industries.
Colour: Black
Certificações: EN ISO 20345:2011; S2, SRC.