Print…
Flame-retardant jackets, BizFlame™ Plus, FR55
Fornecedor: Portwest
FR55NARS FR55NARXXL FR55NARXXXL FR55NARM FR55NARL FR55NARXL
PRTWFR55NARSEA 96.8 EUR
PRTWFR55NARS PRTWFR55NARXXL PRTWFR55NARXXXL PRTWFR55NARM PRTWFR55NARL PRTWFR55NARXL
Flame-retardant jackets, BizFlame™ Plus, FR55
Uniformes
This jacket is constructed from 99% cotton interwoven with 1% carbon fibres. The fabric has flame-resistant and anti-static properties and is therefore ideal for offshore and welding industries.
  • High visibility reflective tape on arms and shoulders for enhanced visibility
  • Concealed front brass zip, triple stitched seams
  • Two front chest pockets with flap and stud closure, two lower pockets with stud closure
  • Adjustable sleeve opening (velcro adjustment)
Weight: 350 g/m²
Certificações: EN ISO 11612 (A1+A2, B1, C1, F1), EN ISO 11611 (A1+A2, Class 2), EN 1149-5