Print…
Glove liners, BO500W
Fornecedor: Showa
BO500-S BO500-M BO500-L BO500-XL BO500-XXL
112-5984EA 48.6 EUR
112-5984 112-5983 112-5982 112-5985 112-5986
Glove liners, BO500W
Luvas
These seamless knitted nylon gloves with elasticated cuffs feature a PU palm coating. The coating protects the hand from oils and abrasion. Ideal for light assembly tasks where a good grip is needed.
  • Lightweight, elastic, low-linting
  • Anatomical shape, resistant to deformation
  • Breathable back of the hand reduces perspiration
  • Minimal allergy risk, no irritation by seams
Suitable industries include agriculture, logistics and construction.
Colour: Black
EN 388 (3131); Cat. II
Certificações: EN 388:2016 (3131X); Cat. II