Print…
Engraving pen with diamond tip
Fornecedor: OUTILS RUBIS
DS4DI DS3 DS3DI DS2F DS1F
811-0102EA 35.9 EUR
811-0102 811-0110 811-0111 OUTIDS2F OUTIDS1F
Engraving pen with diamond tip
Canetas Ink Pens
Hexagonal aluminium scribing pen, 8 mm, with replaceable diamond tip.
  • Refill tip
  • Tip diameter 1,9 mm