Print…
ESD occupational shoes, clogs, Reflexor®, 36903/36913
Fornecedor: Abeba
36903/36 36903/37 36903/39 36903/40 36903/41 36903/42
ABEB36903/36EA 132 EUR
ABEB36903/36 ABEB36903/37 ABEB36903/39 ABEB36903/40 ABEB36903/41 ABEB36903/42
ESD occupational shoes, clogs, Reflexor®, 36903/36913
Sapatos e botas
ESD clog made from smooth leather, respiration absorbing lining, a foldable and adjustable heel strap, and a fuel resistant PU sole.
  • Two replaceable Reflexor® integrated insoles (a massage integrated insole and a smooth comfort integrated insole)
  • Orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Extremely light, slip resistant PU sole
  • Instep area with elastic band
  • Buckle fastening
Colour: White or black
Colour: White or black
Certificações: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; OB, A, E, SRB