Print…
Online SPE products, HyperSep™ Retain™ CX
Fornecedor: Thermo Fisher Scientific
60310-301 60310-302 60311-301 60311-302 60312-301 60312-302
HYPE60310-301EA 484 EUR
HYPE60310-301 HYPE60310-302 HYPE60311-301 HYPE60311-302 HYPE60312-301 HYPE60312-302
Online SPE products, HyperSep™ Retain™ CX
Cromatografia, Colunas
HyperSep™ Retain™-CX for retention of basic and non-polar analytes in a sample matrix when used with an appropriate HPLC column.
  • Effective removal of contaminants such as proteins from samples
  • Compatible with conventional HPLC systems
  • Fast and effective clean-up and concentration of target compounds