Print…
Hose connectors, Nalgene™
Fornecedor: Thermo Fisher Scientific
7210-0500 7211-0500
NALG7210-0500EA 149 EUR
NALG7210-0500 NALG7211-0500
Hose connectors, Nalgene™
Conexões e Conectores
PP, white.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable
Mini hose barb sanitary adapter fitting for carboys and tanks.
Certificações: Biologically compatible when tested according to the U.S. Pharmacopoeia XXII