Print…
Caps for storage tubes, 2D barcoded, Matrix®
Fornecedor: Thermo Fisher Scientific
Matrix®
479-2048EA 175 EUR
479-2048 479-2047 479-2049 479-1338 479-2051 479-2050 479-2046
Caps for storage tubes, 2D barcoded, Matrix®
Tampas Tampas para Tubos de Ensaio
Simplify sample storage with these color-coded caps. Made from Virgin class VI medical grade PP with virgin class VI medical grade silicone O-ring.
  • Free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins
  • Gasketed caps ensure critical sample protection over the storage lifetime
  • Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
  • ScrewTop caps are compatible with storage down to −176 °C