Print…
Container for Hazardous Waste
Fornecedor: HAMILTON ROBOTICS
281520 199201 185319
736-0845EA 38 EUR
736-0845 736-0847 736-0846
Container for Hazardous Waste
Caixas Caixas para Eliminação de Resíduos
The box is used with waste chute 736-0847 and 736-0846.
  • Container complete with yellow PE bag, thickness of foil 120 µm
  • Two types of bags for container available
  • Bags are highly resistant to tearing and puncturing