Print…

Phenolphthalin ≥98.0% (por HPLC, análise de titulação) indicador redox

Fornecedor: TCI

P0095-25G P0095-500G
TCIAP0095-25GEA 37 EUR
TCIAP0095-25G TCIAP0095-500G
Phenolphthalin ≥98.0% (por HPLC, análise de titulação) indicador redox
Phenolphthalin
Fórmula: C₂₀H₁₆O₄
Número MDL : MFCD00036135
Número CAS: 81-90-3
EINECS: 201-384-4
Merck Index: 14,07244
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fcf2d00f39bc6546c5b79baaa08573b6d7f6449&vid=b4fbea5e7b6ddd9367a29f6c824046a449d1611a&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in EtOH (95vol%) almost transparency
Sensitiveness Colorimetric Test : Turn Purple with Copper ion and turn Red purple by adding Hydrogen peroxide


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR