Print…

Phosphotungstic acid sodium salt, Sigma-Aldrich®

Fornecedor: Merck

P6395-100G P6395-500G
SIALP6395-100GEA 314 EUR
SIALP6395-100G SIALP6395-500G
Phosphotungstic acid sodium salt, Sigma-Aldrich®
Phosphotungstic acid sodium salt
Fórmula: Na₃O₄₀PW₁₂
Número CAS: 51312-42-6
EINECS: 257-132-9

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR