Print…
Cut resistant gloves, SivaCut® 830
Fornecedor: KCL by Honeywell
830.10 830.7 830.8 830.9 830.11
KCHL830.10EA 581 EUR
KCHL830.10 KCHL830.7 KCHL830.8 KCHL830.9 KCHL830.11
Cut resistant gloves, SivaCut® 830
Luvas
This glove consists of a mixture of extremely cut resistant fibres (natural latex, para-aramid, glass, steel) and therefore provides cut protection at the highest level (level 5). The shrink-roughened natural latex coating protects the palm as well as the finger knuckles and the back of the hand.
  • Good grip, especially when handling wet parts
  • Very good flexibility and high wearing comfort thanks to knitted cuffs
  • Good puncture resistance regarding metal (not syringes) and glass
  • Prevents user fatigue
Ideal for people working in the glass and beverage industry, in laboratories, or in the waste disposal industry.
Colour: Blue/grey
EN 388 (2543), Cat. II
Certificações: EN 388 (2543), Cat. II