Print…
Cleanroom shirts with long sleeves, 7R014
Fornecedor: FRISTADS KANSAS
100640520L 100640520M 100640520S 100640520XL 100640520XS 1006405202XL 1006405203XL
KWIN100640520LEA 44.5 EUR
KWIN100640520L KWIN100640520M KWIN100640520S KWIN100640520XL KWIN100640520XS KWIN1006405202XL KWIN1006405203XL
Cleanroom shirts with long sleeves, 7R014
Uniformes
These longsleeves are made of XA80 fabric consisting of 100% filament polyester with a moisture transport system. The structure is reminiscent of fine silk.
  • Round neck
  • Moisture-wicking
  • Suitable as underwear
Tests have shown that the material also functions as a pre-filter. Complete underwear worn in combination with cleanroom workwear boosts particle filtration by up to 50%.
Weight: 99 g/m²