Print…
Cleanroom overalls, 8R011
Fornecedor: FRISTADS KANSAS
1006365402XL 1006365403XL 100636540L 100636540M 100636540S 100636540XL 100636540XS
KWIN1006365402XLEA 122.5 EUR
KWIN1006365402XL KWIN1006365403XL KWIN100636540L KWIN100636540M KWIN100636540S KWIN100636540XL KWIN100636540XS
Cleanroom overalls, 8R011
Uniformes
These overalls are made of XA32 fabric consisting of 99% polyester and 1% conductive fibres. The fibres are continuous filaament fibres, which means that the garments do not emit particles or fibres.
  • Concealed zip and snap fasteners
  • Crew neck with concealed snap fastener
  • Elasticated cuffs, Raglan sleeves
  • Open side legs
  • Elasticated waistband with button holes
The carbon filaments produce good static discharge, thus preventing static charging and minimising triboelectric attraction of particles and fibres. This makes the garments very suitable for activities such as vehicle spraying.
Suitable for use in ISO class 7 cleanrooms.
Weight: 125 g/m²