Print…
Safety ankle boots, lace-up, Steelite™ WORK Kumo, FW23
Fornecedor: Portwest
FW23BKR39 FW23BKR47 FW23BKR46 FW23BKR45 FW23BKR44 FW23BKR43 FW23BKR42 FW23BKR40 FW23BKR38 FW23BKR41 FW23BKR48 FW23BKR49 FW23BKR36 FW23BKR37 FW23BKR50
PRTWFW23BKR39EA 48 EUR
PRTWFW23BKR39 PRTWFW23BKR47 PRTWFW23BKR46 PRTWFW23BKR45 PRTWFW23BKR44 PRTWFW23BKR43 PRTWFW23BKR42 PRTWFW23BKR40 PRTWFW23BKR38 PRTWFW23BKR41 PRTWFW23BKR48 PRTWFW23BKR49 PRTWFW23BKR36 PRTWFW23BKR37 PRTWFW23BKR50
Safety ankle boots, lace-up, Steelite™ WORK Kumo, FW23
Sapatos e botas
Anti-static safety ankle boot made from water resistant full grain leather plus an oil resistant, dual-density PU/PU sole.
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Certificações: EN ISO 20345:2011; S3, SRC.