Print…
Safety ankle boots, lace-up, Steelite™ WORK Welted FW17
Fornecedor: Portwest
FW17BRR41 FW17HOR45 FW17HOR44 FW17HOR43 FW17HOR42 FW17HOR41 FW17HOR40 FW17HOR39 FW17BRR47 FW17BRR46 FW17BRR45 FW17BRR42 FW17BRR44 FW17HOR47 FW17BRR40 FW17BRR39 FW17BRR43 FW17HOR46
PRTWFW17BRR41EA 77.3 EUR
PRTWFW17BRR41 PRTWFW17HOR45 PRTWFW17HOR44 PRTWFW17HOR43 PRTWFW17HOR42 PRTWFW17HOR41 PRTWFW17HOR40 PRTWFW17HOR39 PRTWFW17BRR47 PRTWFW17BRR46 PRTWFW17BRR45 PRTWFW17BRR42 PRTWFW17BRR44 PRTWFW17HOR47 PRTWFW17BRR40 PRTWFW17BRR39 PRTWFW17BRR43 PRTWFW17HOR46
Safety ankle boots, lace-up, Steelite™ WORK Welted FW17
Sapatos e botas
Safety ankle boots manufactured from Crazy Horse and Nubuck leather with padded collar and tongue and a rubber sole resistant to oil and heat (300 °C).
  • Padded collars provide all day long comfort
  • Protective steel toecap (200 J)
Colour: Brown/honey.
Colour: Brown/honey
Certificações: EN ISO 345-1:1993; SB, SRA, HRO.