Print…
UHPLC columns, Pinnacle® DB C8 (USP L7)
Fornecedor: Restek
9413512-700 9413513-700 9413515-700 9413522-700 9413523-700 9413525-700 9413532-700 9413533-700 9413535-700 9413552-700 9413553-700 9413555-700 9413562-700 9413563-700 9413565-700 9413572-700 9413573-700 9413575-700 9413512 9413515 9413522 9413525 9413532 9413535 9413552 9413555 9413562 9413565 9413572 9413575 941350210 941350212 9413212 9413232 9413252 9413312 9413312-700 9413313-700 9413315 9413315-700 941331E 9413332 9413332-700 9413333-700 9413335 9413335-700 941333E 9413352 9413352-700 9413353-700 9413355 9413355-700 941335E 941351E 941352E 941353E 941355E 941356E 941357E
REST9413512-700EA 603 EUR
REST9413512-700 REST9413513-700 REST9413515-700 REST9413522-700 REST9413523-700 REST9413525-700 REST9413532-700 REST9413533-700 REST9413535-700 REST9413552-700 REST9413553-700 REST9413555-700 REST9413562-700 REST9413563-700 REST9413565-700 REST9413572-700 REST9413573-700 REST9413575-700 REST9413512 REST9413515 REST9413522 REST9413525 REST9413532 REST9413535 REST9413552 REST9413555 REST9413562 REST9413565 REST9413572 REST9413575 REST941350210 REST941350212 554-1555 554-1556 554-1557 REST9413312 REST9413312-700 REST9413313-700 REST9413315 REST9413315-700 REST941331E REST9413332 REST9413332-700 REST9413333-700 REST9413335 REST9413335-700 REST941333E REST9413352 REST9413352-700 REST9413353-700 REST9413355 REST9413355-700 REST941335E REST941351E REST941352E REST941353E REST941355E REST941356E REST941357E
UHPLC columns, Pinnacle® DB C8 (USP L7)
Cromatografia, Colunas
Highly base-deactivated spherical silica manufactured by Restek. Monomeric C8 bonding. Similar to Pinnacle® DB C18, but the shorter alkyl chain provides less hydrophobic retention. Less retention can be useful for shortening analysis time, if resolution is adequate. Replaces Hypersil® BDS C8 and Pinnacle® C8 Amine.