Print…
UHPLC columns, Pinnacle® DB Biphenyl (USP L11)
Fornecedor: Restek
9409212 RT-9409352 9409355 9409362 9409365 9409512 9409515 9409522 9409525 9409532 9409535 9409552 9409555 9409562 9409232 9409252 9409312 9409315 9409332 9409335 9409565 9409572 9409575 940950210 940950212 9409312-700 9409313-700 9409315-700 940931E 9409332-700 9409333-700 9409335-700 940933E 9409352-700 9409353-700 9409355-700 940935E 9409362-700 9409363-700 9409365-700 940936E 9409512-700 9409513-700 9409515-700 940951E 9409522-700 9409523-700 9409525-700 940952E 9409532-700 9409533-700 9409535-700 940953E 9409552-700 9409553-700 9409555-700 940955E 9409562-700 9409563-700 9409565-700 940956E 9409572-700 9409573-700 9409575-700 940957E
554-1549EA 916 EUR
554-1549 RESTRT-9409352 REST9409355 REST9409362 REST9409365 REST9409512 REST9409515 REST9409522 REST9409525 REST9409532 REST9409535 REST9409552 REST9409555 REST9409562 554-1550 554-1551 REST9409312 REST9409315 REST9409332 REST9409335 REST9409565 REST9409572 REST9409575 REST940950210 REST940950212 REST9409312-700 REST9409313-700 REST9409315-700 REST940931E REST9409332-700 REST9409333-700 REST9409335-700 REST940933E REST9409352-700 REST9409353-700 REST9409355-700 REST940935E REST9409362-700 REST9409363-700 REST9409365-700 REST940936E REST9409512-700 REST9409513-700 REST9409515-700 REST940951E REST9409522-700 REST9409523-700 REST9409525-700 REST940952E REST9409532-700 REST9409533-700 REST9409535-700 REST940953E REST9409552-700 REST9409553-700 REST9409555-700 REST940955E REST9409562-700 REST9409563-700 REST9409565-700 REST940956E REST9409572-700 REST9409573-700 REST9409575-700 REST940957E
UHPLC columns, Pinnacle® DB Biphenyl (USP L11)
Cromatografia, Colunas
Highly base-deactivated spherical silica manufactured by Restek. Pinnacle® DB Phenyl columns offer alternate selectivity to straight chain hydrocarbon phases, especially for aromatic analytes. Replaces Hypersil® BDS Phenyl and Pinnacle® Phenyl Amine.