Print…
Safety sandals, Velcro® fastening, X-Light, 711031/711036
Fornecedor: Abeba
711031/35 711031/36 711031/37 711031/38 711031/39 711031/40 711031/41 711031/42 711031/43 711031/44 711031/45 711031/46 711031/47 711031/48 711036/35 711036/36 711036/37 711036/38 711036/39 711036/40 711036/41 711036/42 711036/43 711036/44 711036/45 711036/46 711036/47 711036/48
ABEB711031/35EA 98.7 EUR
ABEB711031/35 ABEB711031/36 ABEB711031/37 ABEB711031/38 ABEB711031/39 ABEB711031/40 ABEB711031/41 ABEB711031/42 ABEB711031/43 ABEB711031/44 ABEB711031/45 ABEB711031/46 ABEB711031/47 ABEB711031/48 ABEB711036/35 ABEB711036/36 ABEB711036/37 ABEB711036/38 ABEB711036/39 ABEB711036/40 ABEB711036/41 ABEB711036/42 ABEB711036/43 ABEB711036/44 ABEB711036/45 ABEB711036/46 ABEB711036/47 ABEB711036/48
Safety sandals, Velcro® fastening, X-Light, 711031/711036
Sapatos e botas
Anti-static shoes made from leather lightweight air mesh textile inlays and fuel resistant PU sole.
  • Perspiration-absorbent lining with silver point
  • Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbing system
  • Protective steel toecap (200 J)
  • Instep area with elastic band
  • Double Velcro® fastening
Colour: White or black.
Certificações: DGUV 112-191; EN ISO 20345:2011; S1, SRC.