Print…
General purpose gloves, Vital 117 / Vital 124
Fornecedor: MAPA
34124216 34124217 34124218 34124210 34124109
112-5530EA 16.4 EUR
112-5530 112-5531 112-5532 112-5529 112-5533
General purpose gloves, Vital 117 / Vital 124
Luvas
These liquidproof natural latex gloves feature a cotton flock lining. Ideal for non aggressive environments.
  • Excellent dexterity from the suppleness of natural latex
  • Comfortable cotton flock lining
  • Embossed texture for improved grip
Suitable applications include the assembly of small, oil and grease free parts and general cleaning tasks involving household detergents.
Certificações: EN 388:2016 (0010X), EN ISO 374-1 Type B (KPT), EN 374-5 VIRUS, EN 421; Cat. III