Print…
Di-tert-butyl(methyl)phosphonium tetrafluoroborate, Sigma-Aldrich®
Fornecedor: Merck
Sinônimos: (t-Bu)2PMeHBF4
Nível de qualidade M-Clarity™ = MQ100
643777-1G 643777-5G
SIAL643777-1GEA 117 EUR
SIAL643777-1G SIAL643777-5G
Di-tert-butyl(methyl)phosphonium tetrafluoroborate, Sigma-Aldrich®
Di-tert-butyl(methyl)phosphonium tetrafluoroborate
Fórmula: C₉H₂₂BF₄P
Peso Molecular: 248.05 g/mol
Número MDL : MFCD03840579
Número CAS: 479094-62-7