Print…

Ethyl arachidonate, Sigma-Aldrich®

Fornecedor: Merck


Nível de qualidade M-Clarity™ = MQ100

A9135-100MG A9135-1G
SIALA9135-100MGEA 218 EUR
SIALA9135-100MG SIALA9135-1G
Ethyl arachidonate, Sigma-Aldrich®
Ethyl arachidonate
Fórmula: C₂₂H₃₆O₂
Número MDL : MFCD00038340
Número CAS: 1808-26-0

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR