Print…

Tetrabutylphosphonium hydroxide 40% em água

Fornecedor: TCI
Perigo

T2571-25G T2571-250G
TCIAT2571-25GEA 29 EUR
TCIAT2571-25G TCIAT2571-250G
Tetrabutylphosphonium hydroxide 40% em água
Tetrabutylphosphonium hydroxide
Fórmula: C₁₆H₃₇OP
Peso Molecular: 276.45 g/mol
Número MDL : MFCD00068456
Número CAS: 14518-69-5
EINECS: 238-528-0
UN: 3267
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc484adcbbb84e936a7052ad7dafe52127376b99&vid=46f0dbac9db4dd35f90b69d2c596a0e659025c32&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance Colorless to Almost colorless clear liquid
Concentration (Nonaqueous Titration) 40.0 to 44.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR