Print…

Nonylphenol (mixture of isomers)

Fornecedor: TCI
Perigo

N0300-25ML N0300-500ML
TCIAN0300-25MLEA 17 EUR
TCIAN0300-25ML TCIAN0300-500ML
Nonylphenol (mixture of isomers)
Nonylphenol (mixture of isomers)
Fórmula: C₁₅H₂₄O
Peso Molecular: 220.35 g/mol
Ponto de Inflamação: 150°C
Número MDL : MFCD00002396
Número CAS: 84852-15-3
EINECS: 284-325-5
UN: 3145
ADR: 8 II

Encomendar

Especificações dos Resultados do Teste

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) many component mixture


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR