Print…
Supelco® Organic Reference Standard, Hesperetin
Fornecedor: Merck
H4125-10G H4125-1G
H4125-10GEA 156 EUR
H4125-10G H4125-1G
Supelco® Organic Reference Standard, Hesperetin
Padrões Organic Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7acd8da8c48ef1a173a1073b0ad26f7a4f0b1374&vid=7acd8da8c48ef1a173a1073b0ad26f7a4f0b1374&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61000e3f74e8f269dc6a2862913cc52ff0d9c6a5&vid=61000e3f74e8f269dc6a2862913cc52ff0d9c6a5&prid=web&clid=SAPDAM
Encomendar
ESPECIFICAÇÕES
Synonym(s) 3′,5,7-Trihydroxy-4′-methoxyflavanone; 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4-chromanone
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 69097-99-0
MDL No. MFCD00016956
Molecular weight 302,28
Shipping conditions STD
Storage conditions 2…8 °C