Print…

8-Phenyltheophylline, Sigma-Aldrich®

Fornecedor: Merck

SIALP2278-100MGEA 203 EUR
SIALP2278-100MG SIALP2278-1G
8-Phenyltheophylline, Sigma-Aldrich®
8-Phenyltheophylline
Fórmula: C₁₃H₁₂N₄O₂
Número MDL : MFCD00005582
Número CAS: 961-45-5

Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR