Print…
Bromotrispyrrolidinophosphonium hexafluorophosphate, Sigma-Aldrich®
Fornecedor: Merck
Sinônimos: Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, PyBrOP , Bromotripyrrolidinylphosphonium hexafluorophosphate, Bromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)
18565-1G 18565-25G 18565-5G
SIAL18565-1GEA 38.6 EUR
SIAL18565-1G SIAL18565-25G SIAL18565-5G
Bromotrispyrrolidinophosphonium hexafluorophosphate, Sigma-Aldrich®
Bromotrispyrrolidinophosphonium hexafluorophosphate
Fórmula: C₁₂H₂₄BrF₆N₃P₂
Peso Molecular: 466.2 g/mol
Número MDL : MFCD00077412
Número CAS: 132705-51-2