Print…

Bromoxylenol blue

Fornecedor: TCI

B0998-25G B0998-5G
TCIAB0998-25GEA 178 EUR
TCIAB0998-25G TCIAB0998-5G
Bromoxylenol blue
Bromoxylenol blue
Fórmula: C₂₃H₂₀Br₂O₅S
Número MDL : MFCD00005873
Número CAS: 40070-59-5
EINECS: 254-780-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26559bd2907721e60efac872d7097eaf3e7de415&vid=26559bd2907721e60efac872d7097eaf3e7de415&prid=web&clid=SAPDAM

Encomendar

Especificações dos Resultados do Teste

Appearance White to Brown to Dark purple powder to crystal
Lambda max. 612.0 to 616.0 nm (H2O)
Absorbance min. 0.670 (10mg/L, H2O, 612.0 to 616.0nm)
Transition interval (pH) (yellow)6.0 to 7.6 (blue)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR