D(-)-Luciferin potassium salt
Fornecedor: Promega
Sinônimos:
(S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt, Beetle Luciferin potassium salt, D-Luciferin potassium, Luciferin potassium salt, Firefly luciferin potassium salt, Potassium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate
E1601
E1602
E1603
E1605
PROME16010005EA
117
EUR
PROME16010005
PROME16020050
PROME16030250
PROME1605
D(-)-Luciferin potassium salt
D-(-)-Luciferin potassium salt
Fórmula:
C₁₁H₇KN₂O₃S₂ |
Número MDL :
MFCD00044928 Número CAS: 115144-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...