Print…

p-Terphenyl-[D₁₄] (98% D), Sigma-Aldrich®

Fornecedor: Merck


Nível de qualidade M-Clarity™ = MQ200

364630-100MG 364630-500MG
SIAL364630-100MGEA 68.5 EUR
SIAL364630-100MG SIAL364630-500MG
p-Terphenyl-[D₁₄] (98% D), Sigma-Aldrich®
p-Terphenyl-[D₁₄]
Temperatura de Armazenamento: Ambiente
Número CAS: 1718-51-0
UN: 3077
ADR: 9,III

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR