Print…

Iodoacetic anhydride, Sigma-Aldrich®

Fornecedor: Merck


Nível de qualidade M-Clarity™ = MQ100

284262-1G 284262-250MG
SIAL284262-1GEA 366 EUR
SIAL284262-1G SIAL284262-250MG
Iodoacetic anhydride, Sigma-Aldrich®
Iodoacetic anhydride
Fórmula: C₄H₄I₂O₃
Número MDL : MFCD00001080
Número CAS: 54907-61-8
UN: 3261
ADR: 8 II

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR