Print…
VWR®, Tubos de PCR, 0,2 ml
Fornecedor: VWR Collection
732-0547EA 57.6 EUR
732-0547 732-0548
VWR®, Tubos de PCR, 0,2 ml
Tubos Tubos para PCR
These PCR tubes with caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
  • Certified free from DNase, RNase and human DNA
  • Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.