Print…
VWR® Q Path®, MacroStar II, Cassetes
Fornecedor: VWR Collection
720-2225EA 166 EUR
720-2225 720-2226 720-2229 720-2230 720-2231 720-2232 720-2224 720-2227 720-2228
VWR® Q Path®, MacroStar II, Cassetes
Cassetes histológicas Cassetes para Biópsia
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes in pre-filled dispensers for cassette burners. Pack of 2×1000 cassettes and 1×2000 lids.
  • Large opening for the filtration of fluids
  • Hinge and separate lid
  • With 45° writing angle