Print…
Chemical resistant gloves, Showa 874 / 874R
Fornecedor: Showa
874-07 874-08 874-09 874R-09 874R-10 874R-07 874R-08 874R-11 874-10 874-11
112-5942EA 63.1 EUR
112-5942 112-5943 112-5944 112-5948 112-5949 112-5946 112-5947 112-5950 112-5945 BEST874-11
Chemical resistant gloves, Showa 874 / 874R
Luvas
These thin, light gloves with butyl coating protect the hand from a wide array of chemical hazards. Available with a smooth finish or with a rough finish on the fingertips.
  • Impermeable for working in damp or greasy environments
  • Rolled cuff prevents dirt from entering the glove
  • Without special treatment: Minimal allergy risks
  • Designed for easy movement and continuous wear
  • Wrist well protected
Ideal for applications in automotive and chemical industries, in metallurgy and in the public sector.
Certificações: EN 388:2016 (0010X), EN ISO 374-1 Type B (BIK), EN ISO 374-5 VIRUS; Cat. III