Print…
Amostradores de ar VWR®, SAS Super Isolator
Fornecedor: VWR Collection
710-0943EA 3840 EUR
710-0943 710-0944 710-0977 710-0978 710-0945 710-0946
Amostradores de ar VWR®, SAS Super Isolator
Amostradores Amostradores de ar
The SAS instrument control unit is placed outside and connected to the aspirating head using a simple electrical connection. All possible typical contamination sources from the vacuum sampling system are avoided using the SAS Super Isolator. An interesting application is that several heads can be controlled by a single SAS control unit, providing a practical and economical solution.
  • Flexibilidade operacional, seleção de múltiplos idiomas
  • Grava dia, hora, local e volume de ar amostrado, bem como o nome do utilizador
  • Grande ecrã retroiluminado com menu intuitivo; estado operacional ativo confirmado por LED vermelho intermitente
O amostrador de ar SAS Super Isolator usa a mesma tecnologia do amostrador de ar SAS Super microbial.
Informações de encomenda: For a complete system, please order the sampler you require, a battery charger, and the appropriate head.
Please note: An adapter is required when using heads for Petri dishes (90 mm) with samplers for contact plates (55 mm). Sets including a head for 90 mm Petri dishes and the required adapter are available.