Print…
Gel permeation chromatography columns, PolarGel
Fornecedor: VARIAN
PL1117-1800 PL1117-1830 PL1117-6800 PL1117-6830 PL1417-0800 PL1417-0830
VARIPL1117-1800EA 778 EUR
VARIPL1117-1800 VARIPL1117-1830 VARIPL1117-6800 VARIPL1117-6830 VARIPL1417-0800 VARIPL1417-0830
Gel permeation chromatography columns, PolarGel
Cromatografia, Colunas
The PolarGel range is ideal for use with polar solvents, such as dimethyl formamide (DMF) and dimethyl sulfoxide (DMSO), and for solvent combinations such as tetrahydrofuran with water.
  • Medium polarity surface and high mechanical stability
  • Operate in a wide range of solvents and solvent combinations
These eluents are very useful in GPC/SEC to separate polar materials, such as polar resins, modified polysaccharides or complex polar polymers that are difficult to analyze in traditional SEC solvents, such as tetrahydrofuran alone. PolarGel-L is used for low molecular weight polar polymers and PolarGel-M for high MW polar polymers.
These 'mixed bed' columns have a medium polarity surface and high mechanical stability. They are capable of operating in a wide range of solvents and solvent combinations, greatly enhancing your ability to analyze polar polymers that are not necessarily water soluble.
Informações de encomenda: Available in two resolving ranges, PolarGel-L and PolarGel-M