Print…
ESD occupational shoes, slip-on, X-Light, 7131130/7131135
Fornecedor: Abeba
7131130/35 7131130/36 7131130/37 7131130/38 7131130/39 7131130/40 7131130/41 7131130/42 7131130/43 7131130/44 7131130/45 7131130/46 7131130/47 7131130/48 7131135/35 7131135/36 7131135/37 7131135/38 7131135/39 7131135/40 7131135/41 7131135/42 7131135/43 7131135/44 7131135/45 7131135/46 7131135/47 7131135/48
ABEB7131130/35EA 93.1 EUR
ABEB7131130/35 ABEB7131130/36 ABEB7131130/37 ABEB7131130/38 ABEB7131130/39 ABEB7131130/40 ABEB7131130/41 ABEB7131130/42 ABEB7131130/43 ABEB7131130/44 ABEB7131130/45 ABEB7131130/46 ABEB7131130/47 ABEB7131130/48 ABEB7131135/35 ABEB7131135/36 ABEB7131135/37 ABEB7131135/38 ABEB7131135/39 ABEB7131135/40 ABEB7131135/41 ABEB7131135/42 ABEB7131135/43 ABEB7131135/44 ABEB7131135/45 ABEB7131135/46 ABEB7131135/47 ABEB7131135/48
ESD occupational shoes, slip-on, X-Light, 7131130/7131135
Sapatos e botas
ESD shoe made from smooth leather, a non foldable, adjustable heel strap and a lightweight, fuel resistant PU sole.
  • Perspiration-absorbent special lining with Silver Point
  • Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbent heel
  • Instep area with elastic band
Colour: White or black
Certificações: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; OB, A, E, FO, SRC