Cyclopropanesulphonyl chloride ≥97%
Fornecedor: Thermo Fisher Scientific
|
Perigo
|
Sinônimos:
Cyclopropanesulphonylchloride
H29107.06
H29107.03
H29107.06EA
99.6
EUR
H29107.06
H29107.03
Cyclopropanesulphonyl chloride ≥97%
Cyclopropanesulphonyl chloride
Purity: 97%
CAS No.: 139631-62-2
Molecular Formula: C3H5ClO2S
Formula Weight: 140.59
Storage Temperature: 2°C to 8°C
Flash Point: 96°(205°F)
Refractive Index: 1.477
Boiling Point: 60°/2mm
UN No.: 3265
MDL No.: MFCD01631933
Fórmula:
C₃H₅ClO₂S Peso Molecular: 140.59 g/mol Ponto de ebulição: 60 °C (2 mmHg) Densidade: 1.39 Ponto de Inflamação: 96 °C (205 °F) |
Número MDL :
MFCD01631933 Número CAS: 139631-62-2 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...